Order Entry
Poland
ContactUsLinkComponent
Triethanolamine (trolamine) ≥97%, GPR RECTAPUR®
Triethanolamine (trolamine) ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Triethanolamine (trolamine) ≥97%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %
  • Density (20/4):
    1.120 - 1.130
  • Water:
    Max. 0.5 %

Specification Test Results

Frequently Bought Together