EDTA (ethylenediamine tetraacetic acid)
Catalog # HACH700599
Supplier: Hach
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541