Potassium citrate monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6100-05-6
:
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MDL Number:MFCD00150442
Merck Index:14,7623
Melting Pt:230 °C (dec.)
Density:1.98
:324.41 g/mol
Storage Temperature:Ambient
EINECS:212-755-5
Related Information
Frequently Bought Together