Order Entry
Poland
ContactUsLinkComponent
 
 
Disodium oxalate, Certipur®, Supelco®
  1.02407.0060
 :  Merck
 :  
 
Disodium oxalate, Certipur®, Supelco®
  1.02407.0060
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    60 g
  • Seq:
    3377

 

 

  • Solubility:
    37 g/l (20 °C)
  • Melting Point:
    250 - 270 °C (decomposition)
  • Molar Mass:
    134 g/mol
  • Bulk Density:
    600 kg/m3
  • Density:
    2.27 g/cm3 (20 °C)
  • pH:
    8 (30 g/l, H2O, 20 °C)

 

 

 :
 :134 g/mol
 :13,08723
 :250…270 °C
 :2.27 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :200-550-3
 :GHS07
 :Ambient
 :62-76-0
 :Na₂C₂O₄
 :MFCD00012465