Order Entry
Poland
ContactUsLinkComponent
Disodium oxalate ≥99.5%, powder ACS
Irritant
Disodium oxalate ≥99.5%, powder ACS
Supplier:  MP Biomedicals
Certificates
Irritant
Disodium oxalate ≥99.5%, powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Assay:
    ≥99.5%
  • Insoluble Matter:
    ≤0.005%
  • Loss on Drying:
    ≤0.01%
  • Neutrality:
    Passes
  • Chloride ; :
    ≤0.002%
  • Sulfate:
    ≤0.002%
  • Ammonium:
    ≤0.002%
  • Heavy Metals:
    ≤0.002%
  • Iron:
    ≤0.001%
  • Potassium:
    ≤0.005%
  • Substances Darkened by Hot Sulfuric Acid:
    Passes

Specification Test Results

Frequently Bought Together