Sebacic acid
Catalog # MOLEM86335635
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:202.25 g/mol
Merck Index:12,08558
Boiling Pt:374 °C (1013 hPa)
Melting Pt:127…131 °C
Density:1.21 g/cm³ (20 °C)
Flash Pt:221 °C
Storage Temperature:Ambient
EINECS:203-845-5
REACH:01-2119519212-52
CAS Number:111-20-6
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00004440