Dimethyl glutarate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1119-40-0
:
Formula:C₇H₁₂O₄
MDL Number:MFCD00008468
:160.17 g/mol
Merck Index:13,04486
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–37 °C
Density:1.087 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:214-277-2
Specification Test Results
- Solubility:4.3 g/l (20 °C)
- Melting Point:-37 °C
- Molar Mass:160.16 g/mol
- Boiling Point:210 - 215 °C (1013 hPa)
- Vapor Pressure:0.13 hPa (20 °C)
- Flash Point:107 °C
- Refractive Index:2.5 mm2/s (25 °C)
- Explosion Limit:0.9 - 7.9 %(V)
- Density:1.09 g/cm3 (20 °C)
- Ignition Point:365 °C
Specification Test Results
Frequently Bought Together