
Isoamyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-92-2
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00008946
:130.19 g/mol
Merck Index:12,05125
Boiling Pt:142 °C (1013 hPa)
Melting Pt:–78 °C
Density:0.87 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-662-3
UN:1104
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:25 °C
:GHS02
Frequently Bought Together