
4'-Nitroacetanilide, Sigma-Aldrich®
Catalog # SIAL130648-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:180.16 g/mol
Merck Index:12,06677
Boiling Pt:409 °C (1013 hPa)
Melting Pt:215 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:203-169-0
:GHS07
CAS Number:104-04-1
Formula:C₈H₈N₂O₃
MDL Number:MFCD00007303