Order Entry
Poland
ContactUsLinkComponent
 
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Catalog # 83691.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Catalog # 83691.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.90 %
  • Acidity:
    Max. 0.0005 meq/g
  • Dimethylamine:
    Max. 10 ppm
  • Evaporation residue:
    Max. 10 ppm
  • Water:
    Max. 150 ppm

Specification Test Results

About this item

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–60.5 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (DIN 51755) (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284