Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83634.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83634.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.02 %
  • Transmittance (270 nm):
    Min. 10 %
  • Transmittance (275 nm):
    Min. 50 %
  • Transmittance (290 nm):
    Min. 80 %
  • Transmittance (300 nm):
    Min. 90 %
  • Transmittance (330 nm):
    Min. 98 %
  • Fluorescence (as quinine) (365 nm):
    Max. 1 ppb
  • Conforms to BDH 15297:
    Passes test

Specification Test Results

About this item

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–60,5 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (DIN 51755) (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284