N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
Catalog # 9344-13
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- Assay (HCON(CH₃)₂) (by GC, corrected for water):≥99.8 %
- Appearance:Passes Test
- Color (APHA):≤15
- Residue after Evaporation:≤5 ppm
- Titrable Acid (meq/g):≤0.0005
- Titrable Base (meq/g):≤0.003
- Water (H₂O)(by Karl Fischer titrn):≤400 ppm
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:≤0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 310 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 295 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - Amines (as dimethylamine):≤5 ppm
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284