Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Certificates
undefined
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Certificates
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.75 %
  • Acidity (CH3COOH):
    ≤0.01 %