

Acetyl chloride, Sigma-Aldrich®
Catalog # SIAL320129-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:78.5 g/mol
Merck Index:13,00087
Boiling Pt:50.9 °C (1013 hPa)
Melting Pt:–112 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:5 °C
EINECS:200-865-6
UN:1717
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
CAS Number:75-36-5
Formula:C₂H₃ClO
MDL Number:MFCD00000719