

Acetoin dimer 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23147-57-1
:513-86-0
Formula:C₈H₁₆O₄
MDL Number:MFCD00004521
:176.21 g/mol
Merck Index:13,00065
Boiling Pt:148 °C (1013 hPa)
Melting Pt:90…91 °C
Density:0,998 g/cm³ (20 °C)
Storage Temperature:Lodówka
EINECS:208-174-1
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- GC:≥95 %
- Additional info:Polymerization can be reversed by heating at 120°C
- Specific gravity:(25°C) 0.995 to 1.019
Specification Test Results
Frequently Bought Together