3-Hexadecanone, Sigma-Aldrich®
Catalog # SIALH7504-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:240.43 g/mol
Storage Temperature:Ambient
EINECS:242-572-6
CAS Number:18787-64-9
Formula:C₁₆H₃₂O
MDL Number:MFCD00048458