

2-Chloroethyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6099-88-3
:
Formula:C₃H₄ClNS
MDL Number:MFCD00041125
:121.59 g/mol
Boiling Pt:79…80 °C (13 mmHg)
Density:1.265
Flash Pt:96 °C (205 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together