Order Entry
Poland
ContactUsLinkComponent
Trichlorofluoromethane
Trichlorofluoromethane
Catalog # SIALT2290-5X1G
Supplier:  Merck
CAS Number:  
Trichlorofluoromethane
Catalog # SIALT2290-5X1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :137.37 g/mol
Merck Index:12,09770
Boiling Pt:23.8 °C (1013 hPa)
Melting Pt:–111.1 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-892-3
CAS Number:75-69-4
Formula:CCl₃F