Chloroacetic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-11-8
:
Formula:ClCH₂COOH
MDL Number:MFCD00002683
:94.5 g/mol
Merck Index:13,02129
Boiling Pt:188 °C (1013 hPa)
Melting Pt:56…57 °C
Density:1.4043 g/cm³ (20 °C)
EINECS:201-178-4
UN:1751
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:126 °C
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Frequently Bought Together