


Kwas chlorooctowy 99% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-11-8
:
Formula:ClCH₂COOH
MDL Number:MFCD00002683
:94.5 g/mol
Merck Index:14,2112
Boiling Pt:189 °C
Melting Pt:60…63 °C
Density:1,404
Flash Pt:126 °C (258 °F)
Storage Temperature:Temperatura otoczenia
EINECS:201-178-4
UN:1751
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Frequently Bought Together