Order Entry
Poland
ContactUsLinkComponent
Chloroacetic acid 99+%
Chloroacetic acid 99+%
Certificates
undefined
Chloroacetic acid 99+%
Certificates

Specification Test Results

  • Appearance:
    White Crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    61 to 63 °C
  • Water:
    ≤0.3 %
  • Titration with NaOH:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together