Order Entry
Poland
ContactUsLinkComponent
Chloroacetamide for synthesis, Sigma-Aldrich®
Toxic
Health
Chloroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Chloroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    100 g/l (20 °C)
  • Melting Point:
    115 - 118 °C
  • Molar Mass:
    93.51 g/mol
  • Bulk Density:
    760 kg/m3
  • Boiling Point:
    220 °C
  • Vapor Pressure:
    0.0026 hPa (20 °C)
  • Flash Point:
    170 °C
  • Density:
    (20 °C) no data available
  • pH:
    3.8 - 4.1 (40 g/l, H2O, 20 °C)
  • Saturation concentration:
    (20 °C) Air,no data available
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Frequently Bought Together