


Chloroacetonitrile for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-14-2
:
Formula:C₂H₂ClN
MDL Number:MFCD00001885
Boiling Pt:125 °C (1013 hPa)
Melting Pt:–66 °C
Density:1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1 (3),I
:M-Clarity™ quality level = MQ200
:75.5 g/mol
EINECS:203-467-0
UN:2668
:GHS02
:GHS06
:GHS09
Frequently Bought Together