Order Entry
Poland
ContactUsLinkComponent
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane ≥98%
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane ≥98%
Catalog # A19856.14
CAS Number:  
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane ≥98%
Catalog # A19856.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :276.28 g/mol
EINECS:206-562-5
CAS Number:354-51-8
Formula:BrClFCCBrF₂
MDL Number:MFCD00039316