Order Entry
Poland
Orders LinkContactUsLinkComponent
Potassium sorbate

Potassium sorbate

Glass bottle for solids, 1 kg
Catalog # MOLE39220896-1KG
Irritant
Marketsource
Supplier:   Molekula
Potassium sorbate
Catalog # MOLE39220896-1KG
Supplier:  Molekula
Marketsource
CAS Number:  24634-61-5
Potassium sorbate
Potassium sorbate

Glass bottle for solids, 1 kg

CAS Number:  24634-61-5
Price
820 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
  • CAS Number:24634-61-5
  • Formula:CH₃CH=CHCH=CHCOOK
  • MDL Number:MFCD00016546
  • Molecular Weight:150.22 g/mol
  • Merck Index:13,07756
  • Melting Pt:270 °C
  • Density:1.36 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:246-376-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications