
(R)-(-)-1-Aminoindan, Sigma-Aldrich®
Catalog # SIAL445347-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.19 g/mol
Boiling Pt:96 °C (8 mmHg)
Melting Pt:15 °C
Density:1.038 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Refrigerator
CAS Number:10277-74-4
Formula:C₉H₁₁N
MDL Number:MFCD00216669