(S)-(+)-1,3-Butanediol 98%
:24621-61-2
:
:C₄H₁₀O₂
:MFCD00064278
:90.12 g/mol
:109 °C (14 mmHg)
:1 g/cm³
:121 °C
:Ambient
:246-363-0
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4385 to 1.4405 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:+27° to +33° (20°C, 589 nm) (c=1, C2H5OH)
Frequently Bought Together