(R,R)-(-)-2,5-Hexanediol, Sigma-Aldrich®
Catalog # SIAL52792-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:118.18 g/mol
Storage Temperature:Refrigerator
CAS Number:17299-07-9
Formula:C₆H₁₄O₂
MDL Number:MFCD00142339