L(+)-Glutamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
CAS Number:56-85-9
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
Boiling Pt:355
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
Molecular Weight:146.15 g/mol
Merck Index:13,04484
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-292-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together