
Iron(II) sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7782-63-0
:
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719
:278.02 g/mol
Merck Index:13,04091
Melting Pt:64 °C
Density:1.9 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together