Platinium(IV) oxide hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:52785-06-5
:MFCD00011184
Formula:O₂Pt·H₂O
MDL Number:MFCD00066964
:245.09 g/mol
Merck Index:12,07686
Melting Pt:450 °C
Density:10.2 g/cm³ (20 °C)
EINECS:215-223-0
ADR:5.1,II
:M-Clarity™ quality level = MQ100
UN:1479
Frequently Bought Together