Order Entry
Poland
ContactUsLinkComponent
Platinium(IV) oxide hydrate Pt 79-84%
Platinium(IV) oxide hydrate Pt 79-84%
Certificates
undefined
Platinium(IV) oxide hydrate Pt 79-84%

Specification Test Results

  • Appearance:
    Brown to black Powder
  • Assay:
    79 to 84 % (Pt)
  • Surface area:
    ≥90 m²/g (BET)

Specification Test Results

Related Information

Frequently Bought Together