Iridium(IV) chloride hydrate, Sigma-Aldrich®
Catalog # SIAL516996-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:334.03 g/mol (anhy)
:M-Clarity™ quality level = MQ100
EINECS:233-048-8
CAS Number:207399-11-9
:10025-97-5
Formula:Cl₄Ir·xH₂O
MDL Number:MFCD00150243