


Cadmium(II) sulphate hydrate ≥99.995% (trace metals basis), Sigma-Aldrich®
Catalog # SIAL202924-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:226.49 g/mol
Density:3.08 g/cm³ (20 °C)
UN:2570
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:233-331-6
:GHS06
:GHS08
:GHS09
CAS Number:15244-35-6
Formula:CdO₄S·H₂O
MDL Number:MFCD00151655