Cadmium(II) nitrate tetrahydrate, Sigma-Aldrich®
Catalog # SIAL229520-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
Synonyms: Cadmium nitrate tetrahydrate
:
:308.48 g/mol
EINECS:233-710-6
UN:3087
:GHS07
:GHS09
CAS Number:10022-68-1
Formula:CdN₂O₆·4H₂O
MDL Number:MFCD00149626
Boiling Pt:132
Melting Pt:60
Density:2.5 g/cm³ (20 °C)
ADR:5.1 II
:M-Clarity™ quality level = MQ200