
Erbium trichloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL203211-20G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:20 g
- Seq:3290
Specifications
About this item
:
Melting Pt:168 °C
:M-Clarity™ quality level = MQ100
:381.71 g/mol
EINECS:233-385-0
:GHS07
CAS Number:10025-75-9
Formula:Cl₃Er·6H₂O
MDL Number:MFCD00149693