Order Entry
Poland
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate, Sigma-Aldrich®
Aluminium potassium sulphate dodecahydrate, Sigma-Aldrich®
Catalog # SIALA6435-100G
Supplier:  Merck
CAS Number:  
Aluminium potassium sulphate dodecahydrate, Sigma-Aldrich®
Catalog # SIALA6435-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:233-141-3
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143