EDTA dipotassium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25102-12-9
:
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MDL Number:MFCD00150036
:404.46 g/mol
Merck Index:13,03546
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:217-895-0
Frequently Bought Together