tri-Sodium citrate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
Frequently Bought Together