Order Entry
Poland
ContactUsLinkComponent
Hydrobromic acid 47 - 49% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Irritant
Hydrobromic acid 47 - 49% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Hydrobromic acid 47 - 49% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    47.0 - 49.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.002%
  • Material darkens when exposed to air and light.:
    ≤0.05%
  • ACS - Assay (HBr) (by acidimetry):
    47.0 - 49.0 %
  • ACS - Residue after Ignition:
    ≤ 0.002 %
  • ACS - Chloride (Cl):
    ≤ 0.05 %
  • ACS - Iodide (I):
    ≤ 0.003 %
  • ACS - Phosphate (PO₄):
    ≤ 0.001 %
  • ACS - Sulfate and Sulfite (as SO₄):
    ≤ 0.003 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 1 ppm
  • Trace Impurities - Selenium (Se):
    ≤ 0.01 ppm

Specification Test Results

Related Information

Frequently Bought Together