Order Entry
Poland
ContactUsLinkComponent
Semicarbazide hydrochloride for synthesis, Sigma-Aldrich® | Semicarbazide hydrochloride
Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    175 - 177 °C (decomposition)
  • Molar Mass:
    111.53 g/mol
  • Bulk Density:
    740 kg/m3
  • pH:
    1.5 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together