Hydrazinium sulphate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10034-93-2
:
Formula:NH₂NH₂·H₂SO₄
MDL Number:MFCD00044873
:130.12 g/mol
Merck Index:14,04772
Melting Pt:254 °C
Density:1,37 g/cm³ (20 °C)
EINECS:233-110-4
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Iodimetric:≥99.0 %
- Residue after ignition:≤0.05 %
- Chloride (Cl):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together