Order Entry
Poland
ContactUsLinkComponent
Lysates, PC-12 (rat)
Lysates, PC-12 (rat)
Catalog # ENZOADILYCPC101F
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Lysates, PC-12 (rat)
Catalog # ENZOADILYCPC101F
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    200 µG
  • Single Donor/Pooled:
    Pooled
  • Description:
    Lysate, PC-12, rat, heat shocked, 2 mg/ml
  • Seq:
    03
Specifications
About this item
The PC-12 cell line was derived from a transplantable rat adrenal pheochromocytoma. These cells were maintained in Ham's F12K medium with L-glutamine (2 mM) adjusted to contain 1,5 g/L sodium bicarbonate (82,5%), horse serum (15%) and faetal bovine serum (2,5%).
Liquid lysates in SDS-PAGE sample buffer, pH 6,8, heated for 10 minutes at 70 °C and containing Tris-HCl (40 mM), SDS (1%), DTT (50 mM), glycerol (7,5%), and bromophenol blue (0,003%).