

N,N-Dimethyl-1,4-phenylenediammonium dichloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:536-46-9
:MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991
:209.12 g/mol
Merck Index:13,03280
Melting Pt:212 °C (dec.)
Storage Temperature:Ambient
EINECS:208-635-7
UN:2811
ADR:6.1,II
:GHS06
:GHS07
Related Information
Frequently Bought Together