Diethylamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:660-68-4
:
Formula:C₄H₁₂ClN
MDL Number:MFCD00012499
:109.6 g/mol
Merck Index:13,03138
Boiling Pt:320 °C (1013 hPa)
Melting Pt:220…225 °C
Density:1.0475 g/cm³ (21 °C)
:M-Clarity™ quality level = MQ100
EINECS:211-541-9
Frequently Bought Together