Order Entry
Poland
ContactUsLinkComponent
Diethylamine hydrochloride
Diethylamine hydrochloride
Catalog # MOLEM55321156
Supplier:  Molekula
CAS Number:  
Diethylamine hydrochloride
Catalog # MOLEM55321156
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :109.6 g/mol
Merck Index:13,03138
Boiling Pt:320 °C (1013 hPa)
Melting Pt:220…225 °C
Density:1.0475 g/cm³ (21 °C)
EINECS:211-541-9
CAS Number:660-68-4
Formula:C₄H₁₂ClN
MDL Number:MFCD00012499