Ammonium thiocyanate concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32055.608
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:60 mL
- Seq:2385
Specifications
Specification Test Results
- Titer (20°C):0.0995 - 0.1005 mol/l
Specification Test Results
About this item
:
:76.12 g/mol
:Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428