Order Entry
Poland
ContactUsLinkComponent
Potassium hydroxide 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
 
Potassium hydroxide 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
  1.09921.0001
 :  Merck
 :  
undefined
 
Potassium hydroxide 0.1 mol/l in aqueous solution, Titrisol®, Supelco®
  1.09921.0001
 :  Merck
 :  

 

  • Pack type:
    Fibre case
  • Pk:
    1 Ampoul
  • Seq:
    0501

 

 

  • Solubility:
    (20 °C) soluble
  • Density:
    1.09 g/cm3 (20 °C)
  • pH:
    14 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

 

 :
 :56.11 g/mol
 :8,
 :M-Clarity™ quality level = MQ100
 :215-181-3
 :1814
 :GHS05
 :Ambient
 :1310-58-3
 :KOH
 :MFCD00003553