Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
Ammonia solution 28%, GPR RECTAPUR®
 
undefined
 
 
 
Ammonia solution 28%, GPR RECTAPUR®
 

 

  • Assay:
    28 - 32 %
  • Density (20/4):
    0.880 - 0.910
  • Evaporation residue:
    Max. 0.02 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm

 

Frequently Bought Together