Tetrabutylammonium fluoride ≥75% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:429-41-4
:
Formula:C₁₆H₃₆FN
MDL Number:MFCD00011747
:M-Clarity™ quality level = MQ100
:261.47 g/mol
EINECS:207-057-2
Frequently Bought Together